(S)-Di-tert-butyl-5-oxopyrrolidine-1,2-dicarboxylate ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:91229-91-3
:
Formula:C₁₄H₂₃NO₅
MDL Number:MFCD03094770
:285.3361 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:55.0 to 59.0 deg-C
- Specific rotation [a]20/D:-38.0 to -43.0 deg (C=1,CHCl3)