

cis-1,4-Bis(aminomethyl)cyclohexane ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:10029-09-1
:
Formula:C₈H₁₈N₂
:142.246 g/mol
Boiling Pt:76°C
Melting Pt:–9°C
UN:2922
ADR:8,II
:GHS05
:GHS06
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %