Mono(boc)(PEO)-3-diamine, boc-DOOA ≥97.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:153086-78-3
:
Formula:C₁₁H₂₄N₂O₄
MDL Number:MFCD03788155
:248.3 g/mol
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (HPLC) (CAD):min. 97.0 area%
- Purity (Neutralization titration):min. 95.0 %