
9-([1,1'-Biphenyl]-4-yl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole ≥97.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:1391729-66-0
:
Formula:C₃₀H₂₈BNO₂
MDL Number:MFCD29089348
:445.37 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (with Total Nitrogen):min. 97.0 %
- Melting point:229.0 to 233.0 deg-C