kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
4-Bromothiophene-2-methanol
4-Bromothiophene-2-methanol ≥98.0% (by GC)
4-Bromothiophene-2-methanol ≥98.0% (by GC)
:
TCI
4-Bromothiophene-2-methanol ≥98.0% (by GC)
:
TCI
:
79757-77-0
:
:
C₅H₅BrOS
:
MFCD04115392
:
193.07 g/mol
Appearance:
White to Orange to Green powder to lump
Purity (GC):
min. 98.0 %