Order Entry
Norway
ContactUsLinkComponent
 
5-Bromo-2-methylbenzaldehyde ≥95.0% (by GC)
 :  TCI
 
5-Bromo-2-methylbenzaldehyde ≥95.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Light yellow to Light orange powder to lump
  • Purity (GC):
    min. 95.0 %
  • Melting point:
    45.0 to 49.0 deg-C