9-(4-Bromophenyl)-10-(1-naphthyl)anthracene ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1160506-32-0
:
Formula:C₃₀H₁₉Br
MDL Number:MFCD26400490
:459.39 g/mol
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 98.0 %