
1-(tert-Butoxycarbonyl)guanidine ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:219511-71-4
:
Formula:C₆H₁₃N₃O₂
MDL Number:MFCD07369706
:159.189 g/mol
Melting Pt:168°C
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %