10-Bromo-10'-(2-naphthyl)-9,9'-bianthracene ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1172087-81-8
:
Formula:C₃₈H₂₃Br
MDL Number:MFCD22987563
:559.51 g/mol
Specification Test Results
- Appearance:White to Yellow to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 98.0 %