Order Entry
Norway
ContactUsLinkComponent
 
 
Bikinin ≥98.0% (by HPLC, titration analysis)
  TCIAB4826-25MG
 :  TCI
 :  
undefined
 
Bikinin ≥98.0% (by HPLC, titration analysis)
  TCIAB4826-25MG
 :  TCI
 :  

 

  • Fp:
    25 mg
  • Seq:
    2315

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %

 

 

 :
 :273.09 g/mol
 :144°C
 :GHS07
 :Kjøleskap
 :188011-69-0
 :C₉H₉BrN₂O₃
 :MFCD00116442