Benzo[b]thiophene-2-methanol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:17890-56-1
:
Formula:C₉H₈OS
MDL Number:MFCD02682004
:164.22 g/mol
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:98.0 to 102.0 deg-C