kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
Benzo[b]furan-3-boronic acid
Benzo[b]furan-3-boronic acid (contains varying amounts of Anhydride)
Benzo[b]furan-3-boronic acid (contains varying amounts of Anhydride)
:
TCI
Benzo[b]furan-3-boronic acid (contains varying amounts of Anhydride)
:
TCI
:
317830-83-4
:
:
C₈H₇BO₃
:
MFCD06801687
:
161.95 g/mol
:
GHS07
Appearance:
White to Almost white powder to crystal
Purity (Neutralization titration):
97.0 to 113.0 %
Manual Boron Compounds
Manual Synthetic Organic Chemistry and Materials Chemistry