9-Bromo-10-(2-naphthyl)anthracene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:474688-73-8
:
Formula:C₂₄H₁₅Br
MDL Number:MFCD09832882
:383.29 g/mol
Specification Test Results
- Appearance:White to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:169.0 to 173.0 deg-C