
3,6-Dihydro-2H-pyridine-1-carboxylic acid tert-butyl ester ≥97.0% (by GC)
Katalog # TCIAB4357-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
About this item
:
:183.25 g/mol
Boiling Pt:84°C
:GHS07
CAS Number:85838-94-4
Formula:C₁₀H₁₇NO₂
MDL Number:MFCD04972245