7-Benzyloxy-6-methoxy-3H-quinazolin-4-one ≥98.0% (by HPLC, titration analysis)
: TCI
:179688-01-8
:
:C₁₆H₁₄N₂O₃
:MFCD04115119
:282.30 g/mol
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %