2,6-Bis[(2S,5S)-4,4-diphenyl-1-aza-3-oxabicyclo[3.3.0]octan-2-yl]pyridine ≥90.0% (by HPLC)
Catalog # TCIAB4228-50MG
Supplier: TCI
CAS Number:
Specifications
- Pk:50 mg
- Seq:2370
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 90.0 area%
- Specific rotation [a]20/D:-173 to -183 deg (C=1, CHCl3)
Specification Test Results
About this item
:
:605.78 g/mol
CAS Number:1450841-27-6
Formula:C₄₁H₃₉N₃O₂