Order Entry
Norway
ContactUsLinkComponent
tert-Butyl (3R,5S)-6-hydroxy-3,5-O-isopropylidene-3,5-dihydroxyhexanoate ≥98.0% (by GC)
Irritant
tert-Butyl (3R,5S)-6-hydroxy-3,5-O-isopropylidene-3,5-dihydroxyhexanoate ≥98.0% (by GC)
Supplier:  TCI
Certificates
Irritant
tert-Butyl (3R,5S)-6-hydroxy-3,5-O-isopropylidene-3,5-dihydroxyhexanoate ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 98.0 %
  • Specific rotation [a]20/D:
    -3.0 to -5.0 deg (C=2, MeOH)

Specification Test Results

Related Information