4-(Bromomethyl)benzenesulphonyl chloride ≥95.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:66176-39-4
:
Formula:C₇H₆BrClO₂S
MDL Number:MFCD00156129
:269.55
UN:3261
ADR:8,II
:GHS05
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (Argentmetric Titration):min. 95.0 %
- Melting point:73.0 to 77.0 deg-C