kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
5-Bromobenzofuran
5-Bromobenzofuran ≥97.0% (by GC)
5-Bromobenzofuran ≥97.0% (by GC)
:
TCI
5-Bromobenzofuran ≥97.0% (by GC)
:
TCI
:
23145-07-5
:
:
C₈H₅BrO
:
MFCD03407317
:
197.04 g/mol
:
226°C
:
8°C
:
GHS07
Appearance:
Colorless to Light yellow to Light orange clear liquid
Purity (GC):
min. 97.0 %