
N,N,N',N'-Bis(tetramethylene)-O-(N-succinimidyl)uronium hexafluorophosphate ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:207683-26-9
:
Formula:C₁₃H₂₀F₆N₃O₃P
MDL Number:MFCD00192457
:411.29 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:141.0 to 145.0 deg-C
Specification Test Results
Frequently Bought Together