
(4R,6R)-tert-Butyl-6-(2-aminoethyl)-2,2-dimethyl-1,3-dioxane-4-acetate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:125995-13-3
:
Formula:C₁₄H₂₇NO₄
MDL Number:MFCD07369252
:273.3700 g/mol
:GHS07
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 95.0 %
- Specific rotation [a]20/D:+13.0 to +16.0 deg (C=1, CHCl3)