Order Entry
Norway
ContactUsLinkComponent
 
 
tert-Butyl (4R,6R)-6-cyanomethyl-2,2-dimethyl-1,3-dioxane-4-acetate ≥98.0% (by GC, total nitrogen)
  TCIAB3642-5G
 :  TCI
 :  
 
tert-Butyl (4R,6R)-6-cyanomethyl-2,2-dimethyl-1,3-dioxane-4-acetate ≥98.0% (by GC, total nitrogen)
  TCIAB3642-5G
 :  TCI
 :  

 

  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    67.0 to 71.0 deg-C

 

 

 :
 :269.34 g/mol
 :425-780-3
 :GHS07
 :125971-94-0
 :C₁₄H₂₃NO₄
 :MFCD03093958