
4,4'-Bis(chloromethyl)-2,2'-bipyridyl ≥98.0% (by titrimetric analysis)
Catalog # TCIAB3572-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 95.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
About this item
:
:253.13 g/mol
UN:3261
ADR:8,III
:GHS05
CAS Number:138219-98-4
Formula:C₁₂H₁₀Cl₂N₂
MDL Number:MFCD20265579