tert-Butyl-3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:185099-67-6
:
Formula:C₁₂H₁₉NO₃
MDL Number:MFCD00673779
:225.29 g/mol
:GHS05
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:69.0 to 73.0 deg-C