
[Bicyclo[2.2.1]hept-5-en-2-yl]triethoxysilane (mixture of isomers) ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:18401-43-9
:
Formula:C₁₃H₂₄O₃Si
MDL Number:MFCD00167591
:256.42 g/mol
Boiling Pt:104°C
Flash Pt:98°C
EINECS:242-278-8
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 % (total of isomer)