
N-(tert-Butoxycarbonyl)-2,2-dimethyl-1,3-propanediamine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:292606-35-0
:MFCD11227269
Formula:C₁₀H₂₂N₂O₂
MDL Number:MFCD01076212
:202.30 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:72.0 to 76.0 deg-C