(R)-4-Benzyloxymethyl-2,2-dimethyl-1,3-dioxolane ≥97.0% (by GC)
Catalog # TCIAB2136-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
- Specific rotation [a]20/D:-18.0 to -21.0 deg (neat)
Specification Test Results
About this item
:
:222.28 g/mol
CAS Number:14347-83-2
Formula:C₁₃H₁₈O₃
MDL Number:MFCD00066533