
2,2-Bis[4-(4-aminophenoxy)phenyl]propane ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:13080-86-9
:
Formula:C₂₇H₂₆N₂O₂
MDL Number:MFCD00039152
:410.52 g/mol
EINECS:235-985-8
:GHS07
Specification Test Results
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:128.0 to 131.0 deg-C