


1,3-Bis(isocyanatomethyl)cyclohexane (cis and trans mixture) cis- and trans-mixture ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:38661-72-2
:
Formula:C₁₀H₁₄N₂O₂
MDL Number:MFCD00129947
:194.23 g/mol
Flash Pt:159°C
UN:2927
ADR:6.1,II
:GHS05
:GHS06
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 % (total of isomer)