N-[Bis(methylthio)methylene]-p-toluenesulphonamide ≥97.0% (by HPLC, total nitrogen)
Catalog # TCIAB1389-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (with Total Nitrogen):min. 97.0 %
- Melting point:108.0 to 112.0 deg-C
Specification Test Results
About this item
:
:275.40 g/mol
CAS Number:2651-15-2
Formula:C₁₀H₁₃NO₂S₃
MDL Number:MFCD00144854