Order Entry
Norway
ContactUsLinkComponent
Bansyl chloride 10% in hexane
Flammable
Corrosive
Irritant
Health
Bansyl chloride 10% in hexane
Supplier:  TCI
Certificates
Flammable
Corrosive
Irritant
Health
Bansyl chloride 10% in hexane
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light yellow to Yellow clear liquid
  • Concentration (Argentmetric Titration):
    10.0 to 12.0 w/w%

Specification Test Results

Related Information

Frequently Bought Together