
Bansyl chloride ≥98.0% (by HPLC) N-protecting agent, for peptide research
Supplier: TCI
Certificates
About this item
CAS Number:43040-76-2
:
Formula:C₁₈H₂₄ClNO₂S
MDL Number:MFCD00059134
:353.91 g/mol
Storage Temperature:Refrigerator
UN:3265
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Light yellow to Brown clear liquid to cloudy liquid
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 95.0 %
Specification Test Results
Related Information
Frequently Bought Together