N-(4-Methoxy-2-hydroxybenzylidene)-4-butylaniline ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAB0250-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Amber powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Mesomorphic range:42.0 to 67.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:283.37 g/mol
CAS Number:30633-94-4
Formula:C₁₈H₂₁NO₂
MDL Number:MFCD00059253