
Benactyzine hydrochloride ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAB0053-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:175.0 to 179.0 deg-C
Specification Test Results
About this item
:
:363.88 g/mol
Merck Index:14,01029
EINECS:200-324-4
UN:2811
ADR:6.1,III
:GHS06
CAS Number:57-37-4
Formula:C₂₀H₂₅NO₃
MDL Number:MFCD00012624