Order Entry
Norway
ContactUsLinkComponent
 
 
Br-Mmc (=4-bromomethyl-7-methoxycoumarin) ≥98.0% (by titrimetric analysis) for HPLC labeling
 :  TCI
 
 
Br-Mmc (=4-bromomethyl-7-methoxycoumarin) ≥98.0% (by titrimetric analysis) for HPLC labeling
 :  TCI

 

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (HPLC):
    min. 96.0 area%
  • Purity (Argentmetric Titration):
    min. 98.0 %
  • Melting point:
    210.0 to 220.0 deg-C
  • Suitability for Fatty acid analysis_HPLC:
    Effective as labeling agent for Undecanoic Acid

 

Frequently Bought Together