

Nɑ-(5-Fluoro-2,4-dinitrophenyl)-L-leucinamide ≥98.0% (by HPLC) HPLC labeling reagent for e.e. determination
TCIAA5523-100MG
: TCI
:
- Pk:100 mg
- Seq:2440
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Optical purity (LC):min. 99.0 ee%
- Melting point:168.0 to 172.0 deg-C
- Specific rotation [a]20/D:+3.0 to +7.0 deg (C=1, Acetone)
:
:314.27 g/mol
:1325
:4.1,III
:GHS02
:GHS07
:178065-29-7
:C₁₂H₁₅FN₄O₅
:MFCD03844761