Order Entry
Norway
ContactUsLinkComponent
N-Succinimidyl 4-Nitrophenylacetate ≥98.0% (by total nitrogen basis) for HPLC labeling
Irritant
N-Succinimidyl 4-Nitrophenylacetate ≥98.0% (by total nitrogen basis) for HPLC labeling
Catalog # TCIAA5522-1G
Supplier:  TCI
CAS Number:  
Irritant
N-Succinimidyl 4-Nitrophenylacetate ≥98.0% (by total nitrogen basis) for HPLC labeling
Catalog # TCIAA5522-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    181.0 to 185.0 deg-C
  • Solubility in hot Acetone:
    almost transparency
  • Effect test of HPLC derivatization:
    Effective as labeling agent for Diethylamine.

Specification Test Results

About this item

 :
 :278.22 g/mol
EINECS:268-551-1
 :GHS07
Storage Temperature:Refrigerator
CAS Number:68123-33-1
Formula:C₁₂H₁₀N₂O₆
MDL Number:MFCD00047124