2-Aminoperimidine hydrobromide ≥98.0% (by titrimetric analysis) precipitation reagent for SO₄
Supplier: TCI
Certificates
About this item
CAS Number:40835-96-9
:
Formula:C₁₁H₉BrN₃
MDL Number:MFCD00012749
:264.13 g/mol
EINECS:255-101-4
Specification Test Results
- Appearance:White to Light yellow to Dark green powder to crystal
- Purity (Argentmetric Titration):min. 98.0 %
- Solubility in Methanol:almost transparency
- Suitability for Sulfate-analysis:Instantly produce turbidity by sulfateion (5ppm)
Specification Test Results
Frequently Bought Together