Order Entry
Norway
ContactUsLinkComponent
meso-Tetra(4-carboxyphenyl)porphine ≥97.0% (by HPLC, titration analysis) for spectrophotometry
meso-Tetra(4-carboxyphenyl)porphine ≥97.0% (by HPLC, titration analysis) for spectrophotometry
Supplier:  TCI
Certificates
meso-Tetra(4-carboxyphenyl)porphine ≥97.0% (by HPLC, titration analysis) for spectrophotometry
Supplier:  TCI

Specification Test Results

  • Appearance:
    Dark purple powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (Neutralization titration):
    min. 97.0 %
  • Suitability for HPLC-analysis:
    Calibration curve of the copper complex shows linearity

Specification Test Results

Related Information