Alagebrium chloride ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:341028-37-3
:
Formula:C₁₃H₁₄ClNOS
MDL Number:MFCD09027398
:267.77 g/mol
Merck Index:14,00203
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 96.0 %