2-Amino-5-chloro-4-methylpyridine ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:36936-27-3
:
Formula:C₆H₇ClN₂
MDL Number:MFCD06410759
:142.5900 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:151.0 to 155.0 deg-C