
2-Chloro-3-nitropyridin-4-amine ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2789-25-5
:
Formula:C₅H₄ClN₃O₂
MDL Number:MFCD07368898
:173.5572 g/mol
:GHS07
Specification Test Results
- Appearance:White to Yellow powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %