Order Entry
Norway
ContactUsLinkComponent
 
 
 
2-Amyl-5-methylphenol ≥98.0% (by GC)
 :  TCI
 
 
 
2-Amyl-5-methylphenol ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    Light yellow to Brown clear liquid
  • Purity (GC):
    min. 98.0 %