PD 98059 ≥98.0% (by HPLC)
Catalog # TCIAA2529-10MG
Supplier: TCI
CAS Number:
Specifications
- Pk:10 mg
- Seq:2275
Specifications
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):4.90 to 5.40 %
- Melting point:168.0 to 172.0 deg-C
Specification Test Results
About this item
:
:267.284 g/mol
Storage Temperature:Freezer
CAS Number:167869-21-8
Formula:C₁₆H₁₃NO₃
MDL Number:MFCD00671789