(S)-(-)-2-Amino-3-methyl-1,1-diphenyl-1-butanol ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:78603-95-9
:
Formula:C₁₇H₂₁NO
MDL Number:MFCD01318248
:255.36 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:96.0 to 101.0 deg-C
- Specific rotation [a]20/D:-140.0 to -144.0 deg (C=1、CHCL3)