
4-(1-Adamantyl)phenol ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:29799-07-3
:
Formula:C₁₆H₂₀O
MDL Number:MFCD00168143
:228.34 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:179.0 to 183.0 deg-C
Specification Test Results
Frequently Bought Together