Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(+)-Arabinogalactan
(+)-Arabinogalactan (from larch wood)
(+)-Arabinogalactan (from larch wood)
:
TCI
(+)-Arabinogalactan (from larch wood)
:
TCI
:
9036-66-2
:
:
MFCD00062638
:
232-910-0
Appearance:
White to Light yellow to Light orange powder to crystal
Specific rotation [a]20/D:
+7.0 to +12.0 deg (C=1, H2O)
Manual Bioscience and Analyticalscience
Manual Synthetic Organic Chemistry and Materials Chemistry