2-Amino-2'-methyldiphenyl ether ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:3840-18-4
:
Formula:C₁₃H₁₃NO
MDL Number:MFCD00025167
:199.25 g/mol
Boiling Pt:196°C
EINECS:223-329-3
:GHS07
Specification Test Results
- Appearance:Orange to Brown clear liquid to cloudy liquid
- Purity (GC):min. 97.0 %