Order Entry
Norway
ContactUsLinkComponent
5,6,7,8-Tetrahydronaphthalen-1-amine ≥98.0% (by GC, titration analysis)
Irritant
5,6,7,8-Tetrahydronaphthalen-1-amine ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
Irritant
5,6,7,8-Tetrahydronaphthalen-1-amine ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Brown clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results