

2-Amino-5-(diethylamino)toluene monohydrochloride ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAA0289-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in Water:almost transparency
Specification Test Results
About this item
:
:214.74 g/mol
EINECS:218-130-3
UN:2811
ADR:6.1,III
:GHS06
:GHS09
CAS Number:2051-79-8
Formula:C₁₁H₁₈N₂
MDL Number:MFCD00035466